Di-fmoc-D-α,γ-diaminobutyric acid

Supplier: Matrix Scientific

041483-1G 041483-5G
101851-958EA 436.92 USD
101851-958 101851-960
Di-fmoc-D-α,γ-diaminobutyric acid
Di-fmoc-D-alpha,gamma-diaminobutyric acid

Hazard: Irritant

Formula: C₃₄H₃₀N₂O₆
MDL Number: MFCD02094099
CAS Number: 1217645-10-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR