Boc-L-norvaline

Supplier: Matrix Scientific

Synonyms: Boc-Nva-OH, N-(tert-Butoxycarbonyl)-L-norvaline, (2S)-2-Aminopentanoic acid, N-BOC protected, N-Boc-L-norvaline

041480-1G 041480-5G
101851-946EA 40 USD
101851-946 101851-948
Boc-L-norvaline
Boc-L-norvaline

Hazard: Irritant

Formula: C₁₀H₁₉NO₄
MW: 217.265 g/mol
MDL Number: MFCD00037268
CAS Number: 53308-95-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR