N-(tert-Butoxycarbonyl)-L-propargylglycine ≥95%

Supplier: Matrix Scientific

Synonyms: (S)-2-((tert-Butoxycarbonyl)amino)pent-4-ynoic acid, N-Boc-L-propargylglycine, (S)-N-BOC-Propargylglycine, Boc-propargyl-Gly-OH, (S)-2-(Boc-amino)-4-pentynoic Acid, Boc-Pra-OH, (S)-2-(tert-Butoxycarbonylamino)-4-pentynoic Acid

041479-5G 041479-25G
101851-942EA 176.92 USD
101851-942 101851-944
N-(tert-Butoxycarbonyl)-L-propargylglycine ≥95%
N-(tert-Butoxycarbonyl)-L-propargylglycine

Hazard: Irritant/Refrigerate

Formula: C₁₀H₁₅NO₄
MW: 213.23 g/mol
MDL Number: MFCD01320855
CAS Number: 63039-48-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR