(S)-3-Amino-2-(tert-butoxycarbonylamino)propionic acid

Supplier: Matrix Scientific

Synonyms: 3-Amino-N-Boc-L-alanine, Nalpha-Boc-2,3-diaminopropionic Acid, Boc-Dpr-OH, Boc-Dap-OH, 3-Amino-N-(tert-butoxycarbonyl)-L-alanine

041474-1G 041474-5G
101851-922EA 32.31 USD
101851-922 101851-924
(S)-3-Amino-2-(tert-butoxycarbonylamino)propionic acid
(S)-3-Amino-2-(tert-butoxycarbonylamino)propionic acid

Hazard: Irritant

Formula: C₈H₁₆N₂O₄
MW: 204.23 g/mol
MDL Number: MFCD00236843
CAS Number: 73259-81-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR