Boc-L-2-indanylglycine

Supplier: Matrix Scientific

041472-1G 041472-5G
101851-914EA 384.62 USD
101851-914 101851-916
Boc-L-2-indanylglycine
Boc-L-2-indanylglycine

Hazard: Irritant

Formula: C₁₆H₂₁NO₄
MDL Number: MFCD00671384
CAS Number: 181227-47-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR