Boc-L-Nle-OHl . DCHA

Supplier: Matrix Scientific

Synonyms: Boc-Nle-OH·DCHA, Boc-Nle-OH.DCHA

041471-1G 041471-5G
101851-910EA 50.77 USD
101851-910 101851-912
Boc-L-Nle-OHl . DCHA
Boc-L-Nle-OHl . DCHA

Hazard: Irritant

Formula: C₁₁H₂₁NO₄·C₁₂H₂₃N
MW: 412.62 g/mol
MDL Number: MFCD00038968
CAS Number: 21947-32-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR