Boc-aza-L-leucine
Supplier: Matrix Scientific
Synonyms:
N-(tert-Butoxycarbonyl)-3-(dimethylamino)-L-alanine
041470-500MG
041470-1G
101851-906EA
772.31
USD
101851-906
101851-908
Boc-aza-L-leucine
Boc-aza-L-leucine
Hazard: Irritant
Formula:
C₁₀H₂₀N₂O₄ MW: 232.28 g/mol |
MDL Number:
MFCD02682423 CAS Number: 94778-71-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...