N-(tert-Butoxycarbonyl)-4-aminobutyric acid

Supplier: Matrix Scientific

Synonyms: Boc-GABA-OH, N-Boc-γ-aminobutyric acid, 4-(Boc-amino)butyric Acid, 4-(tert-Butoxycarbonylamino)butyric Acid, N-Boc-4-aminobutyric Acid, N-BOC-gamma-Aminobutyric acid

041468-5G 041468-25G
101851-900EA 44.62 USD
101851-900 102551-284
N-(tert-Butoxycarbonyl)-4-aminobutyric acid
N-(tert-Butoxycarbonyl)-4-aminobutyric acid

Hazard: Irritant

Formula: C₉H₁₇NO₄
MW: 203.24 g/mol
MDL Number: MFCD00037313
CAS Number: 57294-38-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR