Boc-D-propargylglycine . DCHA

Supplier: Matrix Scientific

041467-1G 041467-5G
101851-894EA 476.92 USD
101851-894 101851-896
Boc-D-propargylglycine . DCHA
Boc-D-propargylglycine . DCHA

Hazard: Irritant

Formula: C₂₂H₃₈N₂O₄
MW: 394.56 g/mol
MDL Number: MFCD01860891
CAS Number: 63039-47-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR