Boc-D-norvaline
Supplier: Matrix Scientific
|
Warning
|
Synonyms:
(R)-2-((tert-Butoxycarbonyl)amino)pentanoic acid, Boc-D-Nva-OH
041465-1G
041465-5G
101851-886EA
87.7
USD
101851-886
101851-888
Boc-D-norvaline
Boc-D-norvaline
CAS: 57521-85-4
MDL Number: MFCD00155638
Molecular Formula: C10H19NO4
Molecular Weight: 217,27
Hazard: Irritant
Formula:
C₁₀H₁₉NO₄ MW: 217.265 g/mol |
MDL Number:
MFCD00155638 CAS Number: 57521-85-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...