Boc-DL-β-Aib-OH
Supplier: Matrix Scientific
Synonyms:
3-((tert-Butoxycarbonyl)amino)-2-methylpropanoic acid
041464-1G
041464-5G
101851-882EA
250.77
USD
101851-882
101851-884
Boc-DL-β-Aib-OH
Boc-DL-beta-Aib-OH
Hazard: Irritant
Formula:
C₉H₁₇NO₄ MW: 203.24 g/mol |
MDL Number:
MFCD02094137 CAS Number: 16948-10-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...