Boc-DL-3-aminobutanoic acid

Supplier: Matrix Scientific

Synonyms: 3-((tert-Butoxycarbonyl)amino)butanoic acid

041463-1G 041463-5G
101851-878EA 130.77 USD
101851-878 101851-880
Boc-DL-3-aminobutanoic acid
Boc-DL-3-aminobutanoic acid

Hazard: Irritant

Formula: C₉H₁₇NO₄
MDL Number: MFCD00270338
CAS Number: 52815-19-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR