Boc-aza-DL-leucine

Supplier: Matrix Scientific

Synonyms: 2-((tert-Butoxycarbonyl)amino)-3-(dimethylamino)propanoic acid

041462-1G 041462-5G
101851-874EA 615.38 USD
101851-874 101851-876
Boc-aza-DL-leucine
Boc-aza-DL-leucine

Hazard: Irritant

Formula: C₁₀H₂₀N₂O₄
MW: 232.28 g/mol
MDL Number: MFCD04972270
CAS Number: 851653-36-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR