Boc-DL-(2-methylphenyl)glycine

Supplier: Matrix Scientific

041459-1G 041459-5G
101851-862EA 607.69 USD
101851-862 101851-864
Boc-DL-(2-methylphenyl)glycine
Boc-DL-(2-methylphenyl)glycine

Hazard: Irritant

Formula: C₁₄H₁₉NO₄
MW: 265.31 g/mol
MDL Number: MFCD03789594
CAS Number: 40512-48-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR