Boc-DL-(2-methoxyphenyl)glycine
Supplier: Matrix Scientific
041458-1G
041458-5G
101851-858EA
241.54
USD
101851-858
101851-860
Boc-DL-(2-methoxyphenyl)glycine
Boc-DL-(2-methoxyphenyl)glycine
Hazard: Irritant
Formula:
C₁₄H₁₉NO₅ MW: 281.31 g/mol |
MDL Number:
MFCD06797221 CAS Number: 179417-69-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...