Boc-DL-(2-bromophenyl)glycine

Supplier: Matrix Scientific

041457-1G 041457-5G
101851-854EA 615.38 USD
101851-854 101851-856
Boc-DL-(2-bromophenyl)glycine
Boc-DL-(2-bromophenyl)glycine

Hazard: Irritant

Formula: C₁₃H₁₆BrNO₄
MDL Number: MFCD07388844
CAS Number: 1159503-14-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR