Boc-D-Chg-OH

Supplier: Matrix Scientific

Synonyms: Boc-D-Chg-OH, N-(tert-Butoxycarbonyl)-D-2-cyclohexylglycine, N-Boc-2-cyclohexyl-D-glycine, Boc-cyclohexyl-D-Gly-OH, (R)-2-((tert-Butoxycarbonyl)amino)-2-cyclohexylacetic acid, BOC-D-α-cyclohexylglycine, N-Boc-D-2-cyclohexylglycine

041456-1G 041456-5G
101851-850EA 249.23 USD
101851-850 101851-852
Boc-D-Chg-OH
Boc-D-Chg-OH

Hazard: Irritant

Formula: C₁₃H₂₃NO₄
MW: 257.33 g/mol
MDL Number: MFCD00133629
CAS Number: 70491-05-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR