Boc-D-allyl-Gly-OH dicyclohexylammonium salt ≥95%

Supplier: Matrix Scientific

Synonyms: Boc-D-allylglycine (dicyclohexylammonium) salt, 2-Allyl-N-Boc-D-glycine dicyclohexylamine salt, Boc-D-Gly(Allyl)-OH·DCHA, D-2-Allylglycine, N-BOC protected dicylohexylamine salt, Boc-D-allylglycine.DCHA, Dicyclohexylamine (R)-2-((tert-butoxycarbonyl)amino)pent-4-enoate, (R)-2-(Boc-amino)-4-pentenoic acid (dicyclohexylammonium) salt

041452-1G 041452-5G
101851-834EA 36.92 USD
101851-834 101851-836
Boc-D-allyl-Gly-OH dicyclohexylammonium salt ≥95%
Boc-D-allyl-Gly-OH dicyclohexylammonium salt

Hazard: Irritant

Formula: C₁₀H₁₇NO₄·C₁₂H₂₃N
MW: 396.56 g/mol
MDL Number: MFCD01321012
CAS Number: 221352-64-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR