Boc-D-Nle-OH

Supplier: Matrix Scientific

Synonyms: (R)-2-((tert-Butoxycarbonyl)amino)hexanoic acid

041450-1G 041450-5G
101851-826EA 76.92 USD
101851-826 101851-828
Boc-D-Nle-OH
Boc-D-Nle-OH

Hazard: Irritant

Formula: C₁₁H₂₁NO₄
MW: 231.292 g/mol
MDL Number: MFCD00063168
CAS Number: 55674-63-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR