Boc-D-(3-thienyl)glycine

Supplier: Matrix Scientific

041449-100MG 041449-500MG
101851-822EA 661.54 USD
101851-822 101851-824
Boc-D-(3-thienyl)glycine
Boc-D-(3-thienyl)glycine

Hazard: Irritant

Formula: C₁₁H₁₅NO₄S
MW: 257.31 g/mol
MDL Number: MFCD02682481
CAS Number: 33130-97-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR