Boc-D-(2-thienyl)glycine

Supplier: Matrix Scientific

Synonyms: (R)-2-((tert-Butoxycarbonyl)amino)-2-(thiophen-2-yl)acetic acid

041448-100MG 041448-500MG
101851-818EA 547.69 USD
101851-818 101851-820
Boc-D-(2-thienyl)glycine
Boc-D-(2-thienyl)glycine

Hazard: Irritant

Formula: C₁₁H₁₅NO₄S
MW: 257.31 g/mol
MDL Number: MFCD07371938
CAS Number: 74562-03-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR