(S)-2-((tert-Butoxycarbonyl)amino)-2-cyclopentylacetic acid

Supplier: Matrix Scientific

Synonyms: Boc-Cpg-OH·DCHA

041447-250MG 041447-1G
101851-814EA 136.92 USD
101851-814 101851-816
(S)-2-((tert-Butoxycarbonyl)amino)-2-cyclopentylacetic acid
(S)-2-((tert-Butoxycarbonyl)amino)-2-cyclopentylacetic acid

Hazard: Irritant/Store in Freezer

Formula: C₁₂H₂₁NO₄
MW: 243.3 g/mol
MDL Number: MFCD05663437
CAS Number: 109183-72-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR