(S)-2-((tert-Butoxycarbonyl)amino)-2-cyclopentylacetic acid

Supplier: Matrix Scientific

Synonyms: Boc-Cpg-OH·DCHA

041446-250MG 041446-1G
101851-810EA 436.92 USD
101851-810 101851-812
(S)-2-((tert-Butoxycarbonyl)amino)-2-cyclopentylacetic acid
(S)-2-((tert-Butoxycarbonyl)amino)-2-cyclopentylacetic acid

Hazard: Irritant

Formula: C₁₂H₂₁NO₄
MW: 243.3 g/mol
MDL Number: MFCD05663437
CAS Number: 109183-72-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR