Boc-D-NptGly-OH

Supplier: Matrix Scientific

Synonyms: Boc-β-tBu-D-Ala-OH

041444-250MG 041444-1G
101851-802EA 307.69 USD
101851-802 101851-804
Boc-D-NptGly-OH
Boc-D-NptGly-OH

Hazard: Irritant

Formula: C₁₂H₂₃NO₄
MDL Number: MFCD00798621
CAS Number: 112695-98-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR