Boc-β-cyclopropyl-Ala-OH · DCHA

Supplier: Matrix Scientific

Synonyms: Boc-L-Cyclopropylalanine-DCHA, Boc-L-Cyclopropylalanine dicyclohexylamine salt

041443-500MG 041443-1G
101851-798EA 64.62 USD
101851-798 101851-800
Boc-β-cyclopropyl-Ala-OH · DCHA
Boc-β-cyclopropyl-Ala-OH · DCHA

Hazard: Irritant

Formula: C₁₁H₁₉NO₄·C₁₂H₂₃N
MW: 410.60 g/mol
MDL Number: MFCD03093869
CAS Number: 89483-07-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR