Boc-β-cyclopentyl-DL-alanine

Supplier: Matrix Scientific

041442-100MG 041442-500MG
101851-794EA 498.46 USD
101851-794 101851-796
Boc-β-cyclopentyl-DL-alanine
Boc-beta-cyclopentyl-DL-alanine

Hazard: Irritant

Formula: C₁₃H₂₃NO₄
MW: 257.33 g/mol
MDL Number: MFCD02682458
CAS Number: 401514-71-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR