Boc-β-cyclopenten-1-yl-DL-alanine

Supplier: Matrix Scientific

041441-100MG 041441-500MG
101851-790EA 400 USD
101851-790 101851-792
Boc-β-cyclopenten-1-yl-DL-alanine
Boc-beta-cyclopenten-1-yl-DL-alanine

Hazard: Irritant

Formula: C₁₃H₂₁NO₄
MW: 255.32 g/mol
MDL Number: MFCD02682460
CAS Number: 1219411-02-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR