Boc-D-Cha-OH

Supplier: Matrix Scientific

Synonyms: Boc-β-cyclohexyl-D-Ala-OH

041439-1G 041439-5G
101851-782EA 21.54 USD
101851-782 101851-784
Boc-D-Cha-OH
Boc-D-Cha-OH

Hazard: Irritant

Formula: C₁₄H₂₅NO₄
Melting Pt: 64…67 °C
MDL Number: MFCD00133627
CAS Number: 127095-92-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR