Boc-β-Cyano-Ala-OH

Supplier: Matrix Scientific

Synonyms: Boc-β-cyano-Ala-OH

041438-1G 041438-5G
101851-778EA 115.38 USD
101851-778 101851-780
Boc-β-Cyano-Ala-OH
Boc-Beta-Cyano-Ala-OH

Hazard: Irritant

Formula: C₉H₁₄N₂O₄
MW: 214.221 g/mol
MDL Number: MFCD00237539
CAS Number: 45159-34-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR