Boc-β-chloro-Ala-OH

Supplier: Matrix Scientific

Synonyms: Boc-β-chloro-Ala-OH, (R)-2-((tert-Butoxycarbonyl)amino)-3-chloropropanoic acid, Boc-β-Cl-Ala-OH

041437-1G 041437-5G
101851-774EA 275.38 USD
101851-774 101851-776
Boc-β-chloro-Ala-OH
Boc-beta-chloro-Ala-OH

Hazard: Irritant

Formula: C₈H₁₄ClNO₄
MW: 223.653 g/mol
MDL Number: MFCD00672318
CAS Number: 71404-98-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR