N-Boc-3-(4-pyridyl)-L-alanine ≥95%

Supplier: Matrix Scientific

Synonyms: Boc-Ala(4-pyridyl)-OH, Boc-(4-Pyridyl)-Ala-OH, Boc-4-Pal-OH

041436-1G 041436-5G
101851-770EA 52.31 USD
101851-770 101851-772
N-Boc-3-(4-pyridyl)-L-alanine ≥95%
N-Boc-3-(4-pyridyl)-L-alanine

Hazard: Irritant

Formula: C₁₃H₁₈N₂O₄
MW: 266.297 g/mol
MDL Number: MFCD00672528
CAS Number: 37535-57-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR