N-(tert-Butoxycarbonyl)-3-(3-pyridyl)-L-alanine ≥95%

Supplier: Matrix Scientific

Synonyms: (S)-N-BOC-(3-Pyridyl)alanine, N-Boc-3-(3-pyridyl)-L-alanine, Boc-3-Pal-OH, Boc-3-(3-pyridyl)-Ala-OH

041434-1G 041434-5G
101851-762EA 223.08 USD
101851-762 101851-764
N-(tert-Butoxycarbonyl)-3-(3-pyridyl)-L-alanine ≥95%
N-(tert-Butoxycarbonyl)-3-(3-pyridyl)-L-alanine

Hazard: Irritant

Formula: C₁₃H₁₈N₂O₄
MW: 266.30 g/mol
MDL Number: MFCD00079681
CAS Number: 117142-26-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR