Boc-L-3-benzothienylalanine

Supplier: Matrix Scientific

Synonyms: Boc-β-(3-benzothienyl)-Ala-OH, (S)-3-(Benzo[b]thiophen-3-yl)-2-((tert-butoxycarbonyl)amino)propanoic acid

041432-1G 041432-5G
101851-754EA 233.85 USD
101851-754 101851-756
Boc-L-3-benzothienylalanine
Boc-L-3-benzothienylalanine

Hazard: Irritant

Formula: C₁₆H₁₉NO₄S
MW: 321.391 g/mol
MDL Number: MFCD00237538
CAS Number: 154902-51-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR