Boc-3-(3-benzothienyl)-D-alanine

Supplier: Matrix Scientific

Synonyms: Boc-β-(3-benzothienyl)-D-Ala-OH, (R)-3-(Benzo[b]thiophen-3-yl)-2-((tert-butoxycarbonyl)amino)propanoic acid

041431-1G 041431-5G
101851-750EA 300 USD
101851-750 101851-752
Boc-3-(3-benzothienyl)-D-alanine
Boc-3-(3-benzothienyl)-D-alanine

Hazard: Irritant

Formula: C₁₆H₁₉NO₄S
MW: 321.391 g/mol
MDL Number: MFCD00079679
CAS Number: 111082-76-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR