N-Boc-3-(2-thienyl)-L-alanine

Supplier: Matrix Scientific

Synonyms: Boc-β-(2-thienyl)-Ala-OH, Boc-Thi-OH

041430-1G 041430-5G
101851-746EA 187.69 USD
101851-746 101851-748
N-Boc-3-(2-thienyl)-L-alanine
N-Boc-3-(2-thienyl)-L-alanine

Hazard: Irritant

Formula: C₁₂H₁₇NO₄S
MW: 271.331 g/mol
MDL Number: MFCD00062051
CAS Number: 56675-37-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR