N-(tert-Butoxycarbonyl)-3-(2-naphthyl)-L-alanine

Supplier: Matrix Scientific

Synonyms: N-Boc-3-(2-naphthyl)-L-alanine, (S)-alpha-(Boc-amino)-2-naphthalenepropionic Acid, (S)-alpha-tert-Butoxycarbonylamino-2-naphthalenepropionic Acid, Boc-2-Nal-OH

041426-1G 041426-5G
101851-730EA 124.62 USD
101851-730 101851-732
N-(tert-Butoxycarbonyl)-3-(2-naphthyl)-L-alanine
N-(tert-Butoxycarbonyl)-3-(2-naphthyl)-L-alanine

Hazard: Irritant

Formula: C₁₈H₂₁NO₄
MW: 315.37 g/mol
MDL Number: MFCD00079671
CAS Number: 58438-04-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR