N-(tert-Butoxycarbonyl)-3-(2-naphthyl)-D-alanine

Supplier: Matrix Scientific

Synonyms: (R)-alpha-(Boc-amino)-2-naphthalenepropionic Acid, Boc-D-2-Nal-OH, (R)-alpha-tert-Butoxycarbonylamino-2-naphthalenepropionic Acid, N-Boc-3-(2-naphthyl)-D-alanine

041425-1G 041425-5G
101851-726EA 138.46 USD
101851-726 101851-728
N-(tert-Butoxycarbonyl)-3-(2-naphthyl)-D-alanine
N-(tert-Butoxycarbonyl)-3-(2-naphthyl)-D-alanine

Hazard: Irritant

Formula: C₁₈H₂₁NO₄
MW: 315.37 g/mol
MDL Number: MFCD00076900
CAS Number: 76985-10-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR