Boc-3-(1-naphthyl)-L-alanine
Supplier: Matrix Scientific
Synonyms:
Boc-1-Nal-OH, N-Boc-3-(1-naphthyl)-L-alanine, (S)-2-((tert-Butoxycarbonyl)amino)-3-(naphthalen-1-yl)propanoic acid
041424-1G
041424-5G
101851-722EA
224.62
USD
101851-722
101851-724
Boc-3-(1-naphthyl)-L-alanine
Boc-3-(1-naphthyl)-L-alanine
Hazard: Irritant
Formula:
C₁₈H₂₁NO₄ MW: 315.369 g/mol |
MDL Number:
MFCD00085377 CAS Number: 55447-00-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...