Boc-α-methyl-DL-leucine

Supplier: Matrix Scientific

041422-100MG 041422-5G
101851-714EA 138.46 USD
101851-714 101851-716
Boc-α-methyl-DL-leucine
Boc-alpha-methyl-DL-leucine

Hazard: Irritant

Formula: C₁₂H₂₃NO₄
MW: 245.32 g/mol
MDL Number: MFCD02682476
CAS Number: 97534-52-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR