Boc-α-allyl-L-alanine

Supplier: Matrix Scientific

041420-100MG 041420-500MG
101851-706EA 598.46 USD
101851-706 101851-708
Boc-α-allyl-L-alanine
Boc-alpha-allyl-L-alanine

Hazard: Irritant

Formula: C₁₁H₁₉NO₄
MW: 229.28 g/mol
MDL Number: MFCD02682443
CAS Number: 129786-68-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR