Boc-4-hydroxy-D-phenylglycine
Supplier: Matrix Scientific
Synonyms:
(R)-2-((tert-Butoxycarbonyl)amino)-2-(4-hydroxyphenyl)acetic acid, Boc-D-Phg(4-OH)-OH, Boc-D-4-OH-Phg-OH
041417-1G
041417-5G
101851-694EA
412.31
USD
101851-694
101851-696
Boc-4-hydroxy-D-phenylglycine
Boc-4-hydroxy-D-phenylglycine
Hazard: Irritant
Formula:
C₁₃H₁₇NO₅ MW: 267.28 g/mol |
MDL Number:
MFCD01861312 CAS Number: 27460-85-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...