(S)-N-Boc-4-fluorophenylglycine ≥95%
Supplier: Matrix Scientific
Synonyms:
Boc-4-F-Phg-OH, (S)-2-((tert-Butoxycarbonyl)amino)-2-(4-fluorophenyl)acetic acid
041416-1G
041416-5G
101851-690EA
740
USD
101851-690
101851-692
(S)-N-Boc-4-fluorophenylglycine ≥95%
(S)-N-Boc-4-fluorophenylglycine
Hazard: Irritant
Formula:
C₁₃H₁₆FNO₄ MW: 269.272 g/mol |
MDL Number:
MFCD01320862 CAS Number: 142186-36-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...