Boc-4-fluoro-D-phenylglycine
Supplier: Matrix Scientific
Synonyms:
(R)-N-BOC-4-Fluorophenylglycine
041415-1G
041415-5G
101851-686EA
1046.15
USD
101851-686
101851-688
Boc-4-fluoro-D-phenylglycine
Boc-4-fluoro-D-phenylglycine
Hazard: Irritant
Formula:
C₁₃H₁₆FNO₄ MW: 269.272 g/mol |
CAS Number:
196707-32-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...