Boc-4,5-dehydro-Leu-OH·DCHA

Supplier: Matrix Scientific

Synonyms: Dicyclohexylamine (S)-2-((tert-butoxycarbonyl)amino)-4-methylpent-4-enoate

041414-250MG 041414-1G
101851-682EA 300 USD
101851-682 101851-684
Boc-4,5-dehydro-Leu-OH·DCHA
Boc-4,5-dehydro-Leu-OH·DCHA

Hazard: Irritant

Formula: C₂₃H₄₂N₂O₄
MW: 410.599 g/mol
MDL Number: MFCD00237524
CAS Number: 87720-54-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR