Boc-3-amino-4,4,4-trifluorobutyric acid

Supplier: Matrix Scientific

041411-100MG 041411-250MG 041411-500MG
101851-668EA 350.81 USD
101851-668 101851-670 101851-672
Boc-3-amino-4,4,4-trifluorobutyric acid
Boc-3-amino-4,4,4-trifluorobutyric acid

Hazard: Irritant

Formula: C₉H₁₄F₃NO₄
MW: 257.21 g/mol
MDL Number: MFCD02682448
CAS Number: 1185296-42-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR