Boc-2-fluoro-DL-phenylglycine

Supplier: Matrix Scientific

Synonyms: 2-((tert-Butoxycarbonyl)amino)-2-(2-fluorophenyl)acetic acid

041410-1G 041410-5G
101851-664EA 461.54 USD
101851-664 101851-666
Boc-2-fluoro-DL-phenylglycine
Boc-2-fluoro-DL-phenylglycine

Hazard: Irritant

Formula: C₁₃H₁₆FNO₄
MW: 269.28 g/mol
MDL Number: MFCD02682469
CAS Number: 161330-30-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR