Boc-2-amino-4,4,4-trifluorobutyric acid

Supplier: Matrix Scientific

041409-100MG 041409-500MG
101851-660EA 369.23 USD
101851-660 101851-662
Boc-2-amino-4,4,4-trifluorobutyric acid
Boc-2-amino-4,4,4-trifluorobutyric acid

Hazard: Irritant

Formula: C₉H₁₄F₃NO₄
MDL Number: MFCD02682446
CAS Number: 544479-61-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR