Boc-2,5-dihydro-D-phenylglycine
Supplier: Matrix Scientific
041408-1G
041408-5G
101851-656EA
412.31
USD
101851-656
101851-658
Boc-2,5-dihydro-D-phenylglycine
Boc-2,5-dihydro-D-phenylglycine
Hazard: Irritant
Formula:
C₁₃H₁₉NO₄ |
MDL Number:
MFCD01861319 CAS Number: 57872-59-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...