Boc-2,4-dinitro-DL-phenylglycine

Supplier: Matrix Scientific

041407-100MG 041407-500MG
101851-652EA 275.38 USD
101851-652 101851-654
Boc-2,4-dinitro-DL-phenylglycine
Boc-2,4-dinitro-DL-phenylglycine

Hazard: Irritant

Formula: C₁₃H₁₅N₃O₈
MDL Number: MFCD02682466
CAS Number: 1219168-33-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR