Boc-Dap(Fmoc)-OH ≥95%
Supplier: Matrix Scientific
041406-1G
041406-5G
101851-648EA
67.69
USD
101851-648
101851-650
Boc-Dap(Fmoc)-OH ≥95%
Boc-Dap(Fmoc)-OH
Hazard: Irritant
Formula:
C₂₃H₂₆N₂O₆ MW: 426.469 g/mol |
MDL Number:
MFCD00235897 CAS Number: 122235-70-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...